Anti-TMED2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308994EA 667 EUR
ORIGTA308994
Anti-TMED2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: TMED2 (transmembrane emp24 domain trafficking protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR