Anti-UNC13B Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308991EA 667 EUR
ORIGTA308991
Anti-UNC13B Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:1000-1:10000

Type: Primary
Antigen: UNC13B (unc-13 homolog B (C. elegans))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR