Anti-RASSF2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308983EA 667 EUR
ORIGTA308983
Anti-RASSF2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: RASSF2 (Ras association (RalGDS/AF-6) domain family member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR