Anti-RARG Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308949EA 667 EUR
ORIGTA308949
Anti-RARG Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000

Type: Primary
Antigen: RARG (retinoic acid receptor, gamma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR