Anti-SCNN1A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308766EA 519 EUR
ORIGTA308766
Anti-SCNN1A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions:

Type: Primary
Antigen: SCNN1A (sodium channel, non-voltage-gated 1 alpha subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR