Anti-MCM3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308761EA 642 EUR
ORIGTA308761
Anti-MCM3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:5000-1:20000

Type: Primary
Antigen: MCM3 (minichromosome maintenance complex component 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR