Anti-TEAD4 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308632EA 642 EUR
ORIGTA308632
Anti-TEAD4 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: TEAD4 (TEA domain family member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR