Anti-PLEKHJ1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308583EA 642 EUR
ORIGTA308583
Anti-PLEKHJ1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: PLEKHJ1 (pleckstrin homology domain containing, family J member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR