Anti-RFC4 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308528EA 642 EUR
ORIGTA308528
Anti-RFC4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: RFC4 (replication factor C (activator 1) 4, 37kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR