Anti-FIS1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308514EA 642 EUR
ORIGTA308514
Anti-FIS1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:

Type: Primary
Antigen: FIS1 (fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR