Anti-DBH Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308366EA 642 EUR
ORIGTA308366
Anti-DBH Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:1000-1:10000

Type: Primary
Antigen: DBH (dopamine beta-hydroxylase (dopamine beta-monooxygenase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR