Anti-POLR2H Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308326EA 642 EUR
ORIGTA308326
Anti-POLR2H Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: POLR2H (polymerase (RNA) II (DNA directed) polypeptide H)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR