Anti-CCT8 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308295EA 642 EUR
ORIGTA308295
Anti-CCT8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:1000-1:10000

Type: Primary
Antigen: CCT8 (chaperonin containing TCP1, subunit 8 (theta))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR