Anti-IP6K1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308137EA 642 EUR
ORIGTA308137
Anti-IP6K1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:1000-1:10000

Type: Primary
Antigen: IP6K1 (inositol hexakisphosphate kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR