Anti-UGT1A9 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308133EA 655 EUR
ORIGTA308133
Anti-UGT1A9 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:10000

Type: Primary
Antigen: UGT1A9 (UDP glucuronosyltransferase 1 family, polypeptide A9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR