Anti-PCBP2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA308051EA 642 EUR
ORIGTA308051
Anti-PCBP2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: PCBP2 (poly(rC) binding protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR