Anti-IFNA8 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA307940EA 642 EUR
ORIGTA307940
Anti-IFNA8 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: IFNA8 (interferon, alpha 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR