Anti-ALDH1A2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA307922EA 642 EUR
ORIGTA307922
Anti-ALDH1A2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot:1:1000-1:10000

Type: Primary
Antigen: ALDH1A2 (aldehyde dehydrogenase 1 family, member A2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR