Anti-MVD Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA307889EA 655 EUR
ORIGTA307889
Anti-MVD Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence:1:100-1:1000; Western Blot:1:500-1:3000

Type: Primary
Antigen: MVD (mevalonate (diphospho) decarboxylase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR