Anti-FANCD2 Rabbit Monoclonal Antibody [clone: EPR2278(2)]

Lieferant: OriGene
ORIGTA307855EA 669 EUR
ORIGTA307855
Anti-FANCD2 Rabbit Monoclonal Antibody [clone: EPR2278(2)]
Antikörper
Recommended Dilutions: Immunochemistry/Immunofluorescence: 1:200; Western Blot: 1:1000-1:10000

Type: Primary
Antigen: FANCD2 (Fanconi anemia, complementation group D2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR