Anti-TTR Rabbit Monoclonal Antibody [clone: EPR2928(2)]

Lieferant: OriGene
ORIGTA307716EA 786 EUR
ORIGTA307716
Anti-TTR Rabbit Monoclonal Antibody [clone: EPR2928(2)]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1:10-1:100; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: TTR (transthyretin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR