Anti-ANPEP Rabbit Monoclonal Antibody [clone: EPR4058]

Lieferant: OriGene
ORIGTA307568EA 819 EUR
ORIGTA307568
Anti-ANPEP Rabbit Monoclonal Antibody [clone: EPR4058]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1:10-1:100; Immunochemistry: 1:250-1:500

Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR