Anti-ANXA5 Rabbit Monoclonal Antibody [clone: EPR3979]

Lieferant: OriGene
ORIGTA307565EA 810 EUR
ORIGTA307565
Anti-ANXA5 Rabbit Monoclonal Antibody [clone: EPR3979]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: ANXA5 (annexin A5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR