Anti-JAG2 Rabbit Monoclonal Antibody [clone: EPR3646]

Lieferant: OriGene
ORIGTA307463EA 810 EUR
ORIGTA307463
Anti-JAG2 Rabbit Monoclonal Antibody [clone: EPR3646]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: JAG2 (jagged 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR