Anti-ROR2 Rabbit Monoclonal Antibody [clone: EPR3779]

Lieferant: OriGene
ORIGTA307281EA 844 EUR
ORIGTA307281
Anti-ROR2 Rabbit Monoclonal Antibody [clone: EPR3779]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:100

Type: Primary
Antigen: ROR2 (receptor tyrosine kinase-like orphan receptor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR