Anti-PIP5K1C Rabbit Monoclonal Antibody [clone: MAO-R1]

Lieferant: OriGene
ORIGTA307118EA 668 EUR
ORIGTA307118
Anti-PIP5K1C Rabbit Monoclonal Antibody [clone: MAO-R1]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1:10-1:100

Type: Primary
Antigen: PIP5K1C (phosphatidylinositol-4-phosphate 5-kinase, type I, gamma)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR