Anti-CITED4 Rabbit monoclonal antibody [clone: EPR2702]

Lieferant: OriGene
ORIGTA307013EA 669 EUR
ORIGTA307013
Anti-CITED4 Rabbit monoclonal antibody [clone: EPR2702]
Antikörper
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: CITED4 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR