Anti-DCAF12 Chicken Polyclonal Antibody

Lieferant: OriGene
ORIGTA306965EA 588 EUR
ORIGTA306965
Anti-DCAF12 Chicken Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: DCAF12 (DDB1 and CUL4 associated factor 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR