Anti-RSRC1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306925EA 588 EUR
ORIGTA306925
Anti-RSRC1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: RSRC1 (arginine/serine-rich coiled-coil 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR