Anti-CCDC88A Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306896EA 600 EUR
ORIGTA306896
Anti-CCDC88A Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: CCDC88A (coiled-coil domain containing 88A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR