Anti-SQSTM1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306861EA 600 EUR
ORIGTA306861
Anti-SQSTM1 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1-2 μg/ml

Type: Primary
Antigen: SQSTM1 (sequestosome 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR