Anti-ANO1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306852EA 588 EUR
ORIGTA306852
Anti-ANO1 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: ANO1 (anoctamin 1, calcium activated chloride channel)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR