Anti-ST3GAL6 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306781EA 588 EUR
ORIGTA306781
Anti-ST3GAL6 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: ST3GAL6 (ST3 beta-galactoside alpha-2,3-sialyltransferase 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR