Anti-MTCH2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306737EA 588 EUR
ORIGTA306737
Anti-MTCH2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: MTCH2 (mitochondrial carrier 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR