Anti-SLC39A10 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306711EA 583 EUR
ORIGTA306711
Anti-SLC39A10 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: SLC39A10 (solute carrier family 39 (zinc transporter), member 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR