Anti-NUP155 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306613EA 588 EUR
ORIGTA306613
Anti-NUP155 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: NUP155 (nucleoporin 155kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR