Anti-SCARB2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306597EA 588 EUR
ORIGTA306597
Anti-SCARB2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: SCARB2 (scavenger receptor class B, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR