Anti-SLC35D1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306576EA 583 EUR
ORIGTA306576
Anti-SLC35D1 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 1-2 μg/ml; Immunochemistry: 5 μg/ml

Type: Primary
Antigen: SLC35D1 (solute carrier family 35 (UDP-GlcA/UDP-GalNAc transporter), member D1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR