Anti-RGPD5 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306567EA 588 EUR
ORIGTA306567
Anti-RGPD5 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: RGPD5 (RANBP2-like and GRIP domain containing 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR