Anti-GPR124 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306489EA 588 EUR
ORIGTA306489
Anti-GPR124 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2 and 4 ?g/ml; Immunochemistry: 5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: GPR124 (G protein-coupled receptor 124)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR