Anti-TRIM25 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306467EA 588 EUR
ORIGTA306467
Anti-TRIM25 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 20 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: TRIM25 (tripartite motif containing 25)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR