Anti-PTPN11 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306351EA 588 EUR
ORIGTA306351
Anti-PTPN11 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: PTPN11 (protein tyrosine phosphatase, non-receptor type 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR