Anti-TNFSF12 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306313EA 600 EUR
ORIGTA306313
Anti-TNFSF12 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: TNFSF12 (tumor necrosis factor (ligand) superfamily, member 12)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR