Anti-IRAK2 Rabbit Polyclonal Antibody

Lieferant: OriGene
TA306248
ORIGTA306248EA 600 EUR
ORIGTA306248
Anti-IRAK2 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: IRAK2 (interleukin-1 receptor-associated kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR