Anti-EIF4EBP1 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306222EA 600 EUR
ORIGTA306222
Anti-EIF4EBP1 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 2.5-10 μg/ml; Immunochemistry: 2 μg/ml

Type: Primary
Antigen: EIF4EBP1 (eukaryotic translation initiation factor 4E binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR