Anti-BCL2L11 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306192EA 588 EUR
ORIGTA306192
Anti-BCL2L11 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2.5-5 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: BCL2L11 (BCL2-like 11 (apoptosis facilitator))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR