Anti-BCL2L13 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306100EA 600 EUR
ORIGTA306100
Anti-BCL2L13 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: BCL2L13 (BCL2-like 13 (apoptosis facilitator))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR