Anti-IL27RA Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306081EA 588 EUR
ORIGTA306081
Anti-IL27RA Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: IL27RA (interleukin 27 receptor, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR