Anti-BACE2 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA306031EA 588 EUR
ORIGTA306031
Anti-BACE2 Rabbit Polyclonal Antibody
Antikörper
  • Recommended Dilutions: Western Blot: 0.5-1 μg/ml

Type: Primary
Antigen: BACE2 (beta-site APP-cleaving enzyme 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR