Anti-CIDEA Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA305987EA 583 EUR
ORIGTA305987
Anti-CIDEA Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CIDEA (cell death-inducing DFFA-like effector a)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR